3',4'-Dihydroxyacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(3,4-Dihydroxyphenyl)ethan-1-oneAcetopyrocatechol

006589-1G
101790-808EA 41.54 USD
101790-808
3',4'-Dihydroxyacetophenone ≥98%
3',4'-Dihydroxyacetophenone

Hazard code(s): Irritant

Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 117…119 °C
MDL Number: MFCD00016439
CAS Number: 1197-09-7

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR