8-Chloroquinaldine ≥99%
Supplier: Matrix Scientific
Synonyms:
8-Chloro-2-methylquinoline
006427-5G
006427-1G
101790-548EA
44.62
USD
101790-548
101790-550
8-Chloroquinaldine ≥99%
8-Chloroquinaldine
Hazard code(s): Irritant
Formula:
C₁₀H₈ClN MW: 177.63 g/mol Melting Pt: 66…68 °C |
MDL Number:
MFCD00051764 CAS Number: 3033-82-7 |
Specification Test Results
Melting point | 66-68°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...