6-Chloroquinaldine ≥99%
Supplier: Matrix Scientific
006426-5G
006426-1G
101790-544EA
24.62
USD
101790-544
101790-546
6-Chloroquinaldine ≥99%
6-Chloroquinaldine
Hazard code(s): Irritant
Formula:
C₁₀H₈ClN Melting Pt: 94…96 °C |
MDL Number:
MFCD00051735 CAS Number: 92-46-6 |
Specification Test Results
Melting point | 94-96°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...