3-Iodotoluene ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 1-Iodo-3-methylbenzene

006422-100G 006422-25G
101790-536EA 21.54 USD
101790-536 101790-538
3-Iodotoluene ≥98%
3-Iodotoluene

Hazard code(s): Irritant, Refrigerate

Formula: C₇H₇I
MW: 218.04 g/mol
Boiling Pt: 80…82 °C (10mm)
Melting Pt: 27…28 °C
Density: 1.698 g/cm³
MDL Number: MFCD00001050
CAS Number: 625-95-6
EINECS: 210-918-5

Order Now

Specification Test Results

Boiling point 80-82°C/10m
Density 1.698
ND20 1.604

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR