4-Iodotoluene ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: 1-Iodo-4-methylbenzene

006420-100G
101790-530EA 60 USD
101790-530
4-Iodotoluene ≥99%
4-Iodotoluene

Hazard code(s): Irritant

Formula: C₇H₇I
MW: 218.04 g/mol
Boiling Pt: 211.5 °C
Melting Pt: 34…36 °C
Density: 1.678 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00001059
CAS Number: 624-31-7
EINECS: 210-841-7

Order Now

Specification Test Results

Boiling point 211.5°C
Melting point 34-36°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR