4-tert-Butylbenzonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(tert-Butyl)benzonitrilep-tert-Butylbenzonitrile

006404-1G
101790-516EA 15.38 USD
101790-516
4-tert-Butylbenzonitrile ≥98%
4-tert-Butylbenzonitrile

Hazard code(s): Irritant-Harmful

Formula: C₁₁H₁₃N
MW: 159.23 g/mol
Boiling Pt: 128…130 °C (15mm)
Density: 0.94 g/cm³
MDL Number: MFCD00075840
CAS Number: 4210-32-6
EINECS: 224-137-2

Order Now

Specification Test Results

Boiling point 128-130°C/1
Density 0.94
ND20 1.517

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR