1-(2-Thenoyl)-3,3,3-trifluoroacetone ≥99%
Supplier: Matrix Scientific
Synonyms:
Thenoyltrifluoroacetone
, 2-Thenoyltrifluoroacetone
, 4,4,4-Trifluoro-1-(2-thienyl)butane-1,3-dione
, (2-Thenoyl)-ω,ω,ω-trifluoroacetone
, (TTA)
, TTFA
006370-100G
006370-25G
101790-432EA
21.54
USD
101790-432
101790-434
1-(2-Thenoyl)-3,3,3-trifluoroacetone ≥99%
1-(2-Thenoyl)-3,3,3-trifluoroacetone
Hazard code(s): Irritant
Formula:
C₈H₅F₃O₂S MW: 222.19 g/mol Boiling Pt: 96…98 °C (8mm) Melting Pt: 40…44 °C Flash Pt: 111 °C |
MDL Number:
MFCD00005445 CAS Number: 326-91-0 EINECS: 206-316-7 |
Specification Test Results
Boiling point | 96-98°C/8mm |
Melting point | 40-44°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...