2-(Methylthio)benzoic acid ≥97%
Supplier: Matrix Scientific
006303-25G
006303-5G
101790-364EA
121.54
USD
101790-364
101790-362
2-(Methylthio)benzoic acid ≥97%
2-(Methylthio)benzoic acid
Hazard code(s): Irritant
Formula:
C₈H₈O₂S MW: 168.22 g/mol Melting Pt: 170…173 °C |
MDL Number:
MFCD00029934 CAS Number: 3724-10-5 |
Specification Test Results
Melting point | 170-173°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...