N-Ethylphthalimide ≥98%

Supplier: Matrix Scientific

006289-25G
101790-334EA 332.31 USD
101790-334
N-Ethylphthalimide ≥98%
N-Ethylphthalimide

  • RTECS: TI5075000
  • TSCA: T

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 79 °C
MDL Number: MFCD00014583
CAS Number: 5022-29-7
EINECS: 225-707-3

Order Now

Specification Test Results

Melting point 76-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR