1-Heptanesulfonic acid sodium salt ≥98%

Supplier: Matrix Scientific

Synonyms: Sodium 1-heptanesulfonateSodium 1-heptanesulphonateSodium heptane-1-sulfonateSodium heptane-1-sulphonate

006218-5G 006218-25G
101790-244EA 126.15 USD
101790-244 101790-242
1-Heptanesulfonic acid sodium salt ≥98%
1-Heptanesulphonic acid sodium salt

Hazard code(s): Irritant

Formula: H₃C(CH₂)₅CH₂SO₃Na
MW: 202.25 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007543
CAS Number: 22767-50-6
EINECS: 245-210-5
https://us.vwr.com/stibo/web/std.lang.all/07/91/2300791.jpg

Order Now

Specification Test Results

Melting point 300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR