2-(Trifluoromethyl)phenothiazine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Trifluoromethyl-10H-phenothiazine2-(Trifluoromethyl)-10H-phenothiazine

006026-5G 006026-25G
101789-944EA 15.38 USD
101789-944 101789-946
2-(Trifluoromethyl)phenothiazine ≥97%
2-(Trifluoromethyl)phenothiazine

Hazard code(s): Irritant

Formula: C₁₃H₈F₃NS
MW: 267.27 g/mol
Melting Pt: 188…190 °C
MDL Number: MFCD00005018
CAS Number: 92-30-8
EINECS: 202-145-7

Order Now

Specification Test Results

Melting point 188-190°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR