4-(Difluoromethoxy)benzaldehyde ≥98%

Supplier: Matrix Scientific

006018-5G
101789-922EA 60 USD
101789-922
4-(Difluoromethoxy)benzaldehyde ≥98%
4-(Difluoromethoxy)benzaldehyde

Hazard code(s): Irritant, Air Sensitive

Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
Boiling Pt: 222 °C
Density: 1.302 g/cm³
MDL Number: MFCD00042252
CAS Number: 73960-07-3
EINECS: 277-653-5

Order Now

Specification Test Results

Boiling point 58-60°C/0.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR