1-(2-Chloroethyl)-4-fluorobenzene ≥97%

Supplier: Matrix Scientific
Warning

005982-1G 005982-5G
101789-878EA 113.85 USD
101789-878 101789-876
1-(2-Chloroethyl)-4-fluorobenzene ≥97%
1-(2-Chloroethyl)-4-fluorobenzene

Hazard code(s): Irritant

Formula: C₈H₈ClF
MW: 158.6 g/mol
Boiling Pt: 98…99 °C (26mm)
MDL Number: MFCD00155000
CAS Number: 332-43-4
EINECS: 206-364-9

Order Now

Specification Test Results

Boiling point 98-99°C/26m
ND20 1.5077

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR