Pentafluorobenzaldehyde ≥98%

Supplier: Matrix Scientific

Synonyms: 2,3,4,5,6-Pentafluorobenzaldehyde

005937-25G
101789-796EA 166.15 USD
101789-796
Pentafluorobenzaldehyde ≥98%
Pentafluorobenzaldehyde

Hazard code(s): Toxic, Refrigerate, Stored under Argon

Formula: C₇HF₅O
Boiling Pt: 164…166 °C
Melting Pt: 20 °C
Density: 1.588 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003303
CAS Number: 653-37-2
EINECS: 211-502-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 164-166°C
Melting point 20°C
Density 1.588
ND20 1.45

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR