4-Fluoro-2,6-dinitrophenol ≥97%

Supplier: Matrix Scientific

005912-5G
101789-742EA 218.46 USD
101789-742
4-Fluoro-2,6-dinitrophenol ≥97%
4-Fluoro-2,6-dinitrophenol

Hazard code(s): Irritant

Formula: C₆H₃FN₂O₅
MW: 202.1 g/mol
Melting Pt: 54…56 °C
MDL Number: MFCD00179333
CAS Number: 364-32-9

Order Now

Specification Test Results

Melting point 54-56°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR