3,5-Bis(trifluoromethyl)-1H-pyrazole ≥97%

Supplier: Matrix Scientific

Synonyms: 3,5-Bis(trifluoromethyl)pyrazole

005835-5G
101789-612EA 192.31 USD
101789-612
3,5-Bis(trifluoromethyl)-1H-pyrazole ≥97%
3,5-Bis(trifluoromethyl)-1H-pyrazole

Hazard code(s): Irritant

Formula: C₅H₂F₆N₂
MW: 204.07 g/mol
Boiling Pt: 148 °C (1013 hPa)
Melting Pt: 71…72 °C
MDL Number: MFCD00153672
CAS Number: 14704-41-7

Order Now

Specification Test Results

Melting point 71-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR