1-Bromoperfluorooctane ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: Perfluorooctyl bromide1-BromoheptadecafluorooctanePerflubron

005789-25G
101789-536EA 87.69 USD
101789-536
1-Bromoperfluorooctane ≥98%
1-Bromoperfluorooctane

Hazard code(s): Irritant

Formula: BrF₂C(CF₂)₆CF₃
MW: 498.96 g/mol
Boiling Pt: 142 °C
Melting Pt: 6 °C
Density: 1.930 g/cm³
MDL Number: MFCD00042082
CAS Number: 423-55-2
EINECS: 207-028-4
Merck Index: 12,07299

Order Now

Specification Test Results

Boiling point 142°C
Density 1.93
ND20 1.305

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR