1-Nitropyrene ≥97%

Supplier: Matrix Scientific

005775-5G 005775-1G
101789-488EA 53.85 USD
101789-488 101789-490
1-Nitropyrene ≥97%
1-Nitropyrene

Hazard code(s): Irritant

Formula: C₁₆H₉NO₂
Melting Pt: 154…157 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004138
CAS Number: 5522-43-0
EINECS: 226-868-2

Order Now

Specification Test Results

Melting point 154-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR