6-Methoxyquinaldine ≥99%

Supplier: Matrix Scientific
Warning

005761-5G
101789-478EA 60 USD
101789-478
6-Methoxyquinaldine ≥99%
6-Methoxyquinaldine

Hazard code(s): Irritant

Formula: C₁₁H₁₁NO
MW: 173.21 g/mol
Boiling Pt: 95…96 °C (0.5 mmHg)
Melting Pt: 62…64 °C
MDL Number: MFCD00006761
CAS Number: 1078-28-0
EINECS: 214-080-1

Order Now

Specification Test Results

Melting point 62-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR