2-Methoxyphenylboronic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Methoxybenzeneboronic acid
, Dihydroxy(2-methoxyphenyl)borane
, (2-Methoxyphenyl)metaboric acid
005717-25G
005717-100G
102663-558EA
423.08
USD
102663-558
101789-426
2-Methoxyphenylboronic acid ≥97%
2-Methoxyphenylboronic acid
Hazard code(s): Irritant, Store in Freezer
Formula:
C₇H₉BO₃ MW: 151.96 g/mol Melting Pt: 105…107 °C |
MDL Number:
MFCD00236047 CAS Number: 5720-06-9 |
Specification Test Results
Melting point | 105-107°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...