4-Isothiocyanatobenzoic acid

Supplier: Matrix Scientific

005701-5G
101789-386EA 409.23 USD
101789-386
4-Isothiocyanatobenzoic acid
4-Isothiocyanatobenzoic acid

Hazard code(s): Irritant, Lachrymator, Refrigerate

Formula: C₈H₅NO₂S
MW: 179.2 g/mol
Melting Pt: 218…220 °C (dec.)
MDL Number: MFCD00041369
CAS Number: 2131-62-6

Order Now

Specification Test Results

Melting point 218-220°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR