4'-Benzyloxyacetophenone ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Benzyloxyacetophenone
005653-25G
005653-100G
101789-288EA
92.31
USD
101789-288
101789-286
4'-Benzyloxyacetophenone ≥98%
4'-Benzyloxyacetophenone
Hazard code(s): Irritant
Formula:
C₁₅H₁₄O₂ MW: 226.27 g/mol Melting Pt: 91…92 °C |
MDL Number:
MFCD00017247 CAS Number: 54696-05-8 |
Specification Test Results
Melting point | 91-92°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...