2'-Benzyloxyacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-[2-(Benzyloxy)phenyl]ethan-1-one

005652-5G
101789-284EA 170.77 USD
101789-284
2'-Benzyloxyacetophenone ≥98%
2'-Benzyloxyacetophenone
Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Melting Pt: 38…40 °C
MDL Number: MFCD00017240
CAS Number: 31165-67-0

Order Now

Specification Test Results

Melting point 38-40°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR