2,6-Dichloropyrazine ≥97%

Supplier: Matrix Scientific
Warning

005608-25G 005608-100G
101789-232EA 58.46 USD
101789-232 101789-230
2,6-Dichloropyrazine ≥97%
2,6-Dichloropyrazine

Hazard code(s): Irritant

Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Boiling Pt: 120…122 °C (40 mmHg)
Melting Pt: 55…58 °C
Density: 1.713 g/cm³ (20 °C)
MDL Number: MFCD00006125
CAS Number: 4774-14-5
EINECS: 225-316-8

Order Now

Specification Test Results

Melting point 55-58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR