3,4-Dichlorophenethylamine ≥98%

Supplier: Matrix Scientific

005557-5G
101789-124EA 75.38 USD
101789-124
3,4-Dichlorophenethylamine ≥98%
3,4-Dichlorophenethylamine

Hazard code(s): Irritant

Formula: C₈H₉Cl₂N
Boiling Pt: 280 °C
Density: 1.28 g/cm³
MDL Number: MFCD00060617
CAS Number: 21581-45-3

Order Now

Specification Test Results

Boiling point 114°C/1mm
Density 1.28

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR