4-(Trifluoromethyl)-2,3,5,6-tetrafluorothiophenol ≥97%

Supplier: Matrix Scientific

005413-5G
101788-900EA 610.77 USD
101788-900
4-(Trifluoromethyl)-2,3,5,6-tetrafluorothiophenol ≥97%
4-(Trifluoromethyl)-2,3,5,6-tetrafluorothiophenol

Hazard code(s): Irritant

Formula: C₇HF₇S
MW: 250.14 g/mol
Density: 1.66 g/cm³
MDL Number: MFCD00191594
CAS Number: 651-84-3

Order Now

Specification Test Results

Density 1.66

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR