3'-(Trifluoromethyl)acetanilide ≥98%
Supplier: Matrix Scientific
005382-100G
005382-25G
101788-804EA
152.31
USD
101788-804
101788-802
3'-(Trifluoromethyl)acetanilide ≥98%
3'-(Trifluoromethyl)acetanilide
Hazard code(s): Irritant
Formula:
C₉H₈F₃NO MW: 203.16 g/mol Boiling Pt: 287 °C (1013 hPa) Melting Pt: 103…106 °C |
MDL Number:
MFCD00000383 CAS Number: 351-36-0 EINECS: 206-512-2 |
Specification Test Results
Melting point | 103-106°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...