3-(Trifluoromethoxy)benzaldehyde ≥97%

Supplier: Matrix Scientific
Warning

005370-25G
101788-772EA 73.85 USD
101788-772
3-(Trifluoromethoxy)benzaldehyde ≥97%
3-(Trifluoromethoxy)benzaldehyde

Hazard code(s): Irritant

Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 83…86 °C (24mm)
Melting Pt: 450 °C
Density: 1.330 g/cm³
MDL Number: MFCD00042406
CAS Number: 52771-21-8

Order Now

Specification Test Results

Boiling point 83-86°C/24m
Density 1.33
ND20 1.454

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR