2-(Trifluoromethoxy)thiophenol ≥97%
Supplier: Matrix Scientific
Synonyms:
2-(Trifluoromethoxy)benzene-1-thiol
, 2-(Trifluoromethoxy)benzenethiol
005354-5G
005354-1G
101788-732EA
192.31
USD
101788-732
101788-734
2-(Trifluoromethoxy)thiophenol ≥97%
2-(Trifluoromethoxy)thiophenol
Hazard code(s): Irritant, Stench
Formula:
C₇H₅F₃OS MW: 194.18 g/mol Boiling Pt: 53…55 °C (15mm) |
MDL Number:
MFCD00052320 CAS Number: 175278-01-0 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 53-55°C/15m |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...