2,3-Difluorobenzonitrile ≥98%
Supplier: Matrix Scientific
005209-5G
005209-25G
101788-466EA
104.62
USD
101788-466
101788-464
2,3-Difluorobenzonitrile ≥98%
2,3-Difluorobenzonitrile
Hazard code(s): Irritant, Irritant-Harmful
Formula:
C₇H₃F₂N MW: 139.1 g/mol Boiling Pt: 189 °C Density: 1.254 g/cm³ |
MDL Number:
MFCD00009976 CAS Number: 21524-39-0 EINECS: 244-418-3 UN: 3276 ADR: 6.1,III |
Specification Test Results
Boiling point | 189°C |
Density | 1.254 |
ND20 | 1.4882 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...