2,4-Difluorobenzonitrile ≥97%

Supplier: Matrix Scientific

005204-25G 005204-100G
101788-450EA 75.38 USD
101788-450 101788-448
2,4-Difluorobenzonitrile ≥97%
2,4-Difluorobenzonitrile

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 80…83 °C (22.5 mmHg)
Melting Pt: 47…49 °C
MDL Number: MFCD00009826
CAS Number: 3939-09-1
EINECS: 223-523-8
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 47-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR