3,4-Difluorobenzoic acid ≥98%

Supplier: Matrix Scientific
Warning

005201-100G
101788-442EA 80 USD
101788-442
3,4-Difluorobenzoic acid ≥98%
3,4-Difluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 122…124 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011672
CAS Number: 455-86-7
EINECS: 207-249-6

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR