2,6-Difluorobenzaldehyde ≥98%
Supplier: Matrix Scientific
005165-100G
005165-5G
101788-356EA
13.85
USD
101788-356
101788-360
2,6-Difluorobenzaldehyde ≥98%
2,6-Difluorobenzaldehyde
Hazard code(s): Irritant
Formula:
C₇H₄F₂O MW: 142.1 g/mol Boiling Pt: 82…84 °C (15mm) Melting Pt: 15…17 °C Density: 1.317 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00010293 CAS Number: 437-81-0 |
Specification Test Results
Boiling point | 82-84°C/15m |
Melting point | 15-17°C |
Density | 1.317 |
ND20 | 1.507 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...