2,5-Difluorobenzaldehyde ≥98%

Supplier: Matrix Scientific
Warning

005164-100G
101788-354EA 138.46 USD
101788-354
2,5-Difluorobenzaldehyde ≥98%
2,5-Difluorobenzaldehyde

Hazard code(s): Irritant

Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 67…69 °C (17mm)
Density: 1.308 g/cm³
MDL Number: MFCD00010327
CAS Number: 2646-90-4
UN: 1989
ADR: 3,III

Order Now

Specification Test Results

Boiling point 67-69°C/17m
Density 1.308
ND20 1.498

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR