3,5-Difluorobenzaldehyde ≥98%

Supplier: Matrix Scientific
Warning

005160-100G
101788-338EA 284.62 USD
101788-338
3,5-Difluorobenzaldehyde ≥98%
3,5-Difluorobenzaldehyde

Hazard code(s): Irritant, Store in Freezer

Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 61…63 °C (43mm)
Melting Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00010329
CAS Number: 32085-88-4
UN: 1989
ADR: 3,III

Order Now

Specification Test Results

ND20 1.493

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR