2',4'-Dichloro-5'-fluoroacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(2,4-Dichloro-5-fluorophenyl)ethanone

005110-25G 005110-100G
101788-264EA 38.46 USD
101788-264 101788-266
2',4'-Dichloro-5'-fluoroacetophenone ≥98%
2',4'-Dichloro-5'-fluoroacetophenone

Hazard code(s): Irritant

Formula: C₈H₅Cl₂FO
MW: 207.03 g/mol
Boiling Pt: 134…135 °C (15mm)
Melting Pt: 33…35 °C
MDL Number: MFCD00077499
CAS Number: 704-10-9

Order Now

Specification Test Results

Boiling point 134-135°C/1
Melting point 33-35°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR