2,6-Difluoro-3-nitrobenzonitrile ≥98%

Supplier: Matrix Scientific

005024-25G 005024-5G
101788-184EA 90.77 USD
101788-184 101788-182
2,6-Difluoro-3-nitrobenzonitrile ≥98%
2,6-Difluoro-3-nitrobenzonitrile

Hazard code(s): Irritant

Formula: C₇H₂F₂N₂O₂
Melting Pt: 50…54 °C
MDL Number: MFCD00042254
CAS Number: 143879-77-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 50-54°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR