4-Bromocumene ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-4-isopropylbenzene4-Bromoisopropylbenzene

004960-100G 004960-25G
101788-090EA 430.77 USD
101788-090 101788-088
4-Bromocumene ≥95%
4-Bromocumene

Hazard code(s): Irritant

Formula: C₉H₁₁Br
MW: 199.09 g/mol
Boiling Pt: 95…96 °C (12mm)
MDL Number: MFCD00039159
CAS Number: 586-61-8
EINECS: 209-577-5
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Boiling point 95-96°C/12m
ND20 1.536

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR