2-Iodothiophene ≥97% stabilized
Supplier: Matrix Scientific
Stabilization:
Stabilised with copper
004923-100G
004923-25G
101788-004EA
106.15
USD
101788-004
101788-002
2-Iodothiophene ≥97% stabilized
2-Iodothiophene
Hazard code(s): Irritant
Formula:
C₄H₃IS MW: 210.04 g/mol Boiling Pt: 73 °C (15mm) Melting Pt: –40 °C Density: 1.902 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00005424 CAS Number: 3437-95-4 EINECS: 222-342-1 |
Specification Test Results
Boiling point | 73°C/15mm |
Melting point | -40°C |
Density | 1.902 |
ND20 | 1.652 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...