Perfluorocyclohexene ≥97%

Supplier: Matrix Scientific

004862-25G 004862-5G
101787-918EA 296.92 USD
101787-918 101787-916
Perfluorocyclohexene ≥97%
Perfluorocyclohexene

Hazard code(s): Irritant

Formula: C₆F₁₀
MW: 262.05 g/mol
Boiling Pt: 53 °C
Melting Pt: –50 °C
MDL Number: MFCD00037153
CAS Number: 355-75-9
EINECS: 206-592-9

Order Now

Specification Test Results

Boiling point 53°C
Melting point -50°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR