2,5-Diiodobenzoic acid ≥98%

Supplier: Matrix Scientific

004836-5G
101787-878EA 126.15 USD
101787-878
2,5-Diiodobenzoic acid ≥98%
2,5-Diiodobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄I₂O₂
MW: 373.92 g/mol
Melting Pt: 182…185 °C (dec.)
MDL Number: MFCD00045801
CAS Number: 14192-12-2

Order Now

Specification Test Results

Melting point 182-185°C (

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR