α-Chloro-4-ethyltoulene ≥98%

Supplier: Matrix Scientific
Danger

Synonyms: 1-(Chloromethyl)-4-ethylbenzeneα-Chloro-p-ethyltoulene4-Ethylbenzyl chloride

004762-5G
101787-782EA 69.23 USD
101787-782
α-Chloro-4-ethyltoulene ≥98%
α-Chloro-4-ethyltoulene

Hazard code(s): Corrosive, Lachrymator

Formula: C₉H₁₁Cl
MW: 154.64 g/mol
Boiling Pt: 100 °C (10mm)
Density: 1.04 g/cm³
MDL Number: MFCD00039357
CAS Number: 1467-05-6
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Boiling point 100°C/10mm
Density 1.04

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR