3-Bromobenzaldehyde ≥97%

Supplier: Matrix Scientific
Warning

004722-500G
101787-742EA 269.23 USD
101787-742
3-Bromobenzaldehyde ≥97%
3-Bromobenzaldehyde

Hazard code(s): Irritant

Formula: C₇H₅BrO
MW: 185.02 g/mol
Boiling Pt: 228…230 °C
Melting Pt: 18…21 °C
Density: 1.580 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00003345
CAS Number: 3132-99-8
EINECS: 221-526-9

Order Now

Specification Test Results

Boiling point 228-230°C
Density 1.587
ND20 1.594

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR