1-Iodo-2-nitrobenzene ≥97%

Supplier: Matrix Scientific

004654-100G
101787-628EA 96.92 USD
101787-628
1-Iodo-2-nitrobenzene ≥97%
1-Iodo-2-nitrobenzene

Hazard code(s): Irritant

Formula: C₆H₄INO₂
MW: 249.01 g/mol
Boiling Pt: 288…289 °C
Melting Pt: 49…51 °C
Density: 1.9186 g/cm³ (75 °C)
MDL Number: MFCD00007088
CAS Number: 609-73-4
EINECS: 210-200-1

Order Now

Specification Test Results

Boiling point 288-289°C
Melting point 49-51°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR