3-Thenoic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
Thiophene-3-carboxylic acid
, 3-Thiophenecarboxylic acid
004626-100G
004626-25G
101787-598EA
213.85
USD
101787-598
101787-596
3-Thenoic acid ≥97%
3-Thenoic acid
Hazard code(s): Irritant
Formula:
C₅H₄O₂S MW: 128.15 g/mol Melting Pt: 140…141 °C |
MDL Number:
MFCD00005467 CAS Number: 88-13-1 EINECS: 201-802-5 Merck Index: 12,09415 |
Specification Test Results
Melting point | 140-141°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...