3,5-Dibromo-1-fluorobenzene ≥98%

Supplier: Matrix Scientific

Synonyms: 1,3-Dibromo-5-fluorobenzene

004557-100G
101787-476EA 73.85 USD
101787-476
3,5-Dibromo-1-fluorobenzene ≥98%
3,5-Dibromo-1-fluorobenzene

Hazard code(s): Irritant

Formula: C₆H₃Br₂F
MW: 253.9 g/mol
Boiling Pt: 204…206 °C
Density: 2.018 g/cm³
MDL Number: MFCD00061119
CAS Number: 1435-51-4

Order Now

Specification Test Results

Boiling point 204-206°C
Density 2.018
ND20 1.577

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR