2,6-Dibromo-4-(trifluoromethoxy)aniline ≥97%
Supplier: Matrix Scientific
004550-25G
004550-5G
101787-462EA
27.69
USD
101787-462
101787-460
2,6-Dibromo-4-(trifluoromethoxy)aniline ≥97%
2,6-Dibromo-4-(trifluoromethoxy)aniline
Hazard code(s): Irritant
Formula:
C₇H₄Br₂F₃NO Melting Pt: 70…72 °C |
MDL Number:
MFCD00153113 CAS Number: 88149-49-9 |
Specification Test Results
Melting point | 70-72°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...