1,4-Dibromo-1,1,2,2-tetrafluorobutane ≥97%

Supplier: Matrix Scientific

004527-25G 004527-100G
101787-394EA 838.46 USD
101787-394 101787-392
1,4-Dibromo-1,1,2,2-tetrafluorobutane ≥97%
1,4-Dibromo-1,1,2,2-tetrafluorobutane

Hazard code(s): Irritant

Formula: BrF₂CCF₂CH₂CH₂Br
Boiling Pt: 72 °C (80mm)
Density: 1.973 g/cm³
MDL Number: MFCD00039224
CAS Number: 18599-20-7
EINECS: 242-438-7

Order Now

Specification Test Results

Boiling point 72°C/80mm
Density 1.973
ND20 1.416

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR