2,4,6-Trimethylpyrylium tetrafluoroborate ≥98%

Supplier: Matrix Scientific

Synonyms: 2,4,6-Trimethylpyryliumtetrafluoroborate

004498-5G
101787-336EA 102.35 USD
101787-336
2,4,6-Trimethylpyrylium tetrafluoroborate ≥98%
2,4,6-Trimethylpyrylium tetrafluoroborate

Hazard code(s): Irritant

Formula: C₈H₁₁BF₄O
MW: 209.98 g/mol
Melting Pt: 224…226 °C
MDL Number: MFCD00032098
CAS Number: 773-01-3
EINECS: 212-256-2

Order Now

Specification Test Results

Melting point 224-226°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR