Trifluoromethanesulfonamide ≥98%

Supplier: Matrix Scientific

004449-1G 004449-5G
101787-266EA 51.75 USD
101787-266 101787-264
Trifluoromethanesulfonamide ≥98%
Trifluoromethanesulphonamide

Hazard code(s): Irritant

Formula: CH₂F₃NO₂S
Melting Pt: 120…122 °C
MDL Number: MFCD00068714
CAS Number: 421-85-2
EINECS: 431-270-1

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR